2-Ethylhexyl acetate ≥98.0%

Supplier: TCI America

A0031-500ML A0031-25ML
TCA0031-500MLEA 248.06 CAD
TCA0031-500ML TCA0031-025ML
2-Ethylhexyl acetate ≥98.0%
2-Ethylhexyl acetate

CAS Number: 103-09-3
MDL Number: MFCD00027249
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Melting point (°C): -80
Flash Point (°C): 77
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Acetic Acid 2-Ethylhexyl Ester, Octyl Acetate

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 200 °C
Melting Pt: –80 °C
Density: 0.87 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00027249
CAS Number: 103-09-3
EINECS: 203-079-1
Merck Index: 12,06860
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c8a786760177d8428ed4c8510e8a1e0efac7062&vid=0c8a786760177d8428ed4c8510e8a1e0efac7062&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR