4'-Methoxyacetanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Methoxyphenyl)acetamide

A0019-25G A0019-100G
TCA0019-025GEA 132.72 CAD
TCA0019-025G TCA0019-100G
4'-Methoxyacetanilide ≥98.0%
4'-Methoxyacetanilide

CAS Number: 51-66-1
MDL Number: MFCD00014963
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 130

Formula: C₉H₁₁NO₂
Melting Pt: 130 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00014963
CAS Number: 51-66-1
EINECS: 200-114-2
Merck Index: 13,00052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ae921914704282bbf06082411d816778404f91e&vid=4ae921914704282bbf06082411d816778404f91e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR