Diethyl acetamidomalonate ≥98.0%
Supplier: TCI America
Synonyms:
Diethyl 2-acetamidomalonate
A0015-25G
A0015-500G
A0015-100G
TCA0015-025GEA
36.34
CAD
TCA0015-025G
TCA0015-500G
TCA0015-100G
Diethyl acetamidomalonate ≥98.0%
Diethyl acetamidomalonate
CAS Number: 1068-90-2
MDL Number: MFCD00009146
Molecular Formula: C9H15NO5
Molecular Weight: 217.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96
Other Product Names / Synonyms: Acetamidomalonic Acid Diethyl Ester, Ethyl Acetamidomalonate
Formula:
C₉H₁₅NO₅ MW: 217.22 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD00009146 CAS Number: 1068-90-2 EINECS: 213-952-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...