4-Acetamidobenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Formylphenyl)acetamide, 4'-Formylacetanilide

A0009-5G A0009-25G
TCA0009-5GEA 60.04 CAD
TCA0009-5G TCA0009-025G
4-Acetamidobenzaldehyde ≥98.0%
4-Acetamidobenzaldehyde

CAS Number: 122-85-0
MDL Number: MFCD00003380
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 156

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00003380
CAS Number: 122-85-0
EINECS: 204-579-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a91373a8a5301863ccb6552aa1f853054cc15e21&vid=a91373a8a5301863ccb6552aa1f853054cc15e21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR