Acetamidine hydrochloride ≥95.0%

Supplier: TCI America

A0008-25G A0008-500G
TCA0008-025GEA 53.72 CAD
TCA0008-025G TCA0008-500G
Acetamidine hydrochloride ≥95.0%
Acetamidine hydrochloride

CAS Number: 124-42-5
MDL Number: MFCD00013016
Molecular Formula: C2H6N2
Molecular Weight: 94.54
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 168

Formula: C₂H₇ClN₂
Melting Pt: 168 °C
MDL Number: MFCD00013016
CAS Number: 124-42-5
EINECS: 204-700-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f36469ce5f1ed8d0e035a896ac2538a85fe2e01&vid=2f36469ce5f1ed8d0e035a896ac2538a85fe2e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR