Acenaphthenequinone ≥98.0%

Supplier: TCI America
Warning

A0004-25G A0004-100G
TCA0004-025GEA 382.36 CAD
TCA0004-025G TCA0004-100G
Acenaphthenequinone ≥98.0%
Acenaphthenequinone

CAS Number: 82-86-0
MDL Number: MFCD00003805
Molecular Formula: C12H6O2
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 255


Other Product Names / Synonyms: Acenaphthaquinone

Formula: C₁₂H₆O₂
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 255 °C
Density: 1.48 g/cm³ (20 °C)
MDL Number: MFCD00003805
CAS Number: 82-86-0
EINECS: 201-441-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4126752b2ef1cd921b4c2dec841582c4ef04a40&vid=b4126752b2ef1cd921b4c2dec841582c4ef04a40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR