Vacuum Stopcock with Easy-Action Plug, Glass, Ace Glass Incorporated
Supplier: Ace Glass
8194-67 8194-69 8194-74 8194-76
89062-548EA 358.43 CAD
89062-548 89062-550 89062-552 89062-554
Vacuum Stopcock with Easy-Action Plug, Glass, Ace Glass Incorporated
Stopcocks
This straight-through stopcock is all borosilicate glass (including the plug), with a nylon handle
Stopcocks 89062-548 and -552 use barrel 89062-480 and O-Rings 89062-562. Stopcocks 89062-550 and -554 use barrel 89062-482 and O-Rings 89062-564.
Pressures up to 10-5 Torr are attainable after baking.
Ordering information: Stopcocks are available complete with plug and barrel, or individual components may be purchased separately. Supplied with FETFE O-Rings.