Vacuum Traps, Threaded with Serrated Hose Connections, Ace Glass
Supplier: Ace Glass
8747-37 8747-11 8747-33 8747-39 8747-43
89063-840EA 489.55 CAD
89063-840 89063-868 89063-870 89063-874 89063-876
Vacuum Traps, Threaded with Serrated Hose Connections, Ace Glass
Vacuum Traps
This vacuum trap consists of a glass body with Ace-Thred that accepts an adjustable inner tube secured by nylon bushing 89059-220 and FETFE O-ring 89060-984
The adjustable inlet and outlet tubes have six-ring hose connections (the O.D. of the smallest ring is 14mm; the largest, 16mm). Body fabricated from borosilicate glass.
Ordering information: Available as a complete item, or individual components may be purchased separately. Complete item includes glass body, glass inner tube 89063-868, and nylon bushing 89059-220 with FETFE O-ring 89060-984.