Clamps, Pinch-Type, Ace Glass
Supplier: Ace Glass
7669-03 7669-08 7669-10 7669-12 7669-14 7669-16 7669-18 7669-20 7669-22 7669-05
89060-250EA 23.25 CAD
89060-250 89060-252 89060-254 89060-256 89060-258 89060-260 89060-262 89060-264 89060-266 89060-268 89060-270
Clamps, Pinch-Type, Ace Glass
Clamps Standard Taper and Spherical Joint Clamps
These all-stainless-steel pinch clamps are for use with o-ring spherical joints, and with ball and socket joints
Sizes 12 and 18 (only) are available either spring loaded or with a screw-locking device. All other sizes have the screw-locking feature.