PTFE Replacement Stopcock Plug, Ace Glass Incorporated
Supplier: Ace Glass
8194-270 8194-266 8194-268 8194-272
89062-438EA 183.59 CAD
89062-438 89062-484 89062-486 89062-492
PTFE Replacement Stopcock Plug, Ace Glass Incorporated
Stopcocks
PTFE replacement stopcock plugs have a UHDPE (Ultra High Density Polyethylene) hood, and each plug includes three FETFE O-Rings
The handle can be removed from the plug, making the plug bakeable.
Accessories information: Tapered plastic sleeve is used to fit O-Rings onto stopcock plugs.
Ordering information: For replacement sets of these O-Rings, order 89062-502 (0–3, 0–4), 89062-504 (0–5, 0–8), 89062-506 (0–10), or 89062-508 (0–15).