High Vacuum Stopcock with Easy-Action Plug, Ace Glass Incorporated
Supplier: Ace Glass
8194-224 8194-226 8194-228 8194-230 8194-232 8194-234 8194-25 8194-26 8194-27 8194-28 8194-35 8194-34
89062-468EA 267.77 CAD
89062-468 89062-470 89062-472 89062-474 89062-476 89062-478 89062-480 89062-482 89062-488 89062-494 89062-512 89062-620
High Vacuum Stopcock with Easy-Action Plug, Ace Glass Incorporated
Stopcocks
These high vacuum stopcocks are for use in applications involving at least 3.5Kg/cm² of internal pressure
Stopcocks 89062-480, -494, -472, and -474 accommodate replacement plug 89062-486 and replacement FETFE O-Ring 89062-562. Stopcocks 89062-620, -512, -468, and -470 accommodate replacement plug 89062-484 and replacement FETFE O-Ring 89062-560. Stopcocks 89062-482 and -476 accommodate replacement plug 89062-438 and replacement FETFE O-Ring 89062-564. Stopcocks 89062-488 and -478 accommodate replacement plug 89062-492 and replacement FETFE O-Ring 89062-566.
They are designed with variable openings from 0–3mm to 0–15mm. An ultimate vacuum of 10-7 can be obtained with the (standard) three O-Rings. A smooth acting semi-needle valve permits fine adjustment of the opening. The front O-Ring makes a positive closure against a precision formed heavy glass seat. The hooded handle can be removed from the PTFE plug, making the plug bakeable. Reference marks on the body aid in repetitive settings.
Ordering information: Available complete with borosilicate glass barrel and PTFE plug with O-Rings, or components may be purchased separately.