Chromatography Apparatus, Neutral Oil, Ace Glass Incorporated
Supplier: Ace Glass
5869-02 5869-06 5869-14 5869-20 5869-40 5869-08
89053-110EA 423.9 CAD
89053-110 89053-112 89053-116 89053-118 89053-120 89057-578
Chromatography Apparatus, Neutral Oil, Ace Glass Incorporated
Chromatography Systems Complete HPLC Systems
This chromatograpy apparatus features a flask that increases the efficiency and standardization of neutral oil determination by: permitting automatic transfer of weighed crude oil samples onto alumina columns; and converting column operation to a semi-automatic level, thereby reducing the cost of operation
The complete apparatus consists of: 175mL reservoir with a 4mm PTFE stopcock, [ST] 19/22 inner joint at bottom and #27 stopper joint at top; 20mL flask with [ST] 19/22 joints at top and bottom and a [ST] 7/15 joint on inner tube; base with [ST] 19/22 joint; extension tube with [ST] 7/15More Product Information