Float, Polyethylene, Ace Glass Incorporated
Supplier: Ace Glass
5849-05 5849-10 5849-15 5849-20
89052-916PK 41.76 CAD
89052-916 89052-918 89052-920 89052-922
Float, Polyethylene, Ace Glass Incorporated
Seals Security Seals
This float is used on top of chromatography column packing to prevent turbulence caused by a drop-wise addition.
Order Now
SPECIFICATIONS
Environmentally Preferable