Three-Layer Flat Septa, Ace Glass Incorporated
Supplier: Ace Glass
12901-42 12901-48
89077-538PK 57.53 CAD
89077-538 89077-540
Three-Layer Flat Septa, Ace Glass Incorporated
Closures Bottle Closures
Three-layer flat septa consists of two harder layers of rubber outside, and a softer, silicone rubber layer inside
Available in either 7 or 10mm O.D. sizes, both of which are 4mm thick. The 89077-538 item is ideal for 89051-212 electrode adapter, while the 89077-540 is great for #7 Ace-Thred bushings such as 89051-120. These septa reseal very easily.