Resin and Reaction Flask Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Supplier: Ace Glass
12058-44 12058-47 12058-49 12058-51 12058-53 12058-55
89076-094EA 2995.73 CAD
89076-094 89076-096 89076-098 89076-100 89076-102 89076-104
Resin and Reaction Flask Mantle, Aluminum Housing, Bottom Opening, Ace Glass Incorporated
Heaters Heating Mantles
Resin and reaction flask mantle is designed mainly for use with pressure reactors with a bottom valve or outlet
Mantle 89076-094 is designed for flasks having a flat bottom with rounded corners, such as 89054-406.
These mantles also fit many other cylindrical reaction flasks, such as 89083-812, 89055-162, 89055-122 and 89083-998 (among others) with appropriate O.D. Mantles have an aluminum housing and a bottom opening. Temperature range is ambient plus 10 to 450°C (842°F).
Certifications: CSA approved.
Ordering information: Mantle includes 122cm (4') detachable cord with lockingMore Product Information