Spinwedge Magnetic Stir Bar, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13665-02 13665-04 13665-06 13665-08
89084-224EA 25.82 CAD
89084-224 89084-226 89084-228 89084-230
Spinwedge Magnetic Stir Bar, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Wedge-shaped PTFE magnetic stir bars are designed to “plow” up sediment for thorough dispersion
Provides very strong magnetic action. Ideal for viscous solutions.