D-(+)-Sucrose ACS, VWR Chemicals BDH®

Supplier: VWR International

BDH9308-500G BDH9308-2.5KG
BDH9308-500GCS 1985.8 CAD
BDH9308-500G BDH9308-2.5KG
D-(+)-Sucrose ACS, VWR Chemicals BDH®
D(+)-Sucrose

White solid

Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.005 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Invert Sugar = 0.05 %
Iron = 0.0005 %
Loss on Drying = 0.03 %
Residue after Ignition = 0.01 %
Specific Rotation +66.3 - +66.8 degrees
Sulfate & Sulfite = 0.005 %
Titrable Free Acid = 0.0008 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR