Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Disodium sulfate, Bisodium sulfate, Disodium sulphate, Sulfuric acid disodium salt, Bisodium sulphate, Sulphuric acid disodium salt

BDH9302-12KG BDH9302-2.5KG BDH9302-500G
BDH9302-12KGEA 574.43 CAD
BDH9302-12KG BDH9302-2.5KG BDH9302-500G
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Sodium sulphate

Meets reagent specifications for testing USP/NF monographs.

Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <= 0.01 %
Chloride <= 0.001 %
Heavy Metals (as Pb) <= 0.0005 %
Insolubles <= 0.01 %
Iron <= 0.001 %
Loss on Ignition <= 0.5 %
Magnesium <= 0.005 %
Nitrogen Compounds <= 0.0005 %
pH (5%, Water) @25C 5.2 - 9.2
Phosphate <= 0.001 %
Potassium <= 0.01 %
Purity >= 99.0 %
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR