Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

BDH9296-500G BDH9296-2.5KG
BDH9296-500GCS 4965.01 CAD
BDH9296-500G BDH9296-2.5KG
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
di-Sodium hydrogen phosphate heptahydrate

Meets reagent specifications for testing USP/NF monographs.


Crystals.

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

Chloride = 0.001 %
Heavy Metals (as Pb) = 0.001 %
Insolubles = 0.005 %
Iron = 0.001 %
pH (5%, Water) @25C 8.7 - 9.3
Purity (as is) 98.0 - 102.0 %
Sulfate = 0.005 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR