Sodium hydroxide ≥97.0%, beads ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
BDH9292-12KG
BDH9292-2.5KG
BDH9292-500G
BDH9292-12KGEA
416.07
CAD
BDH9292-12KG
BDH9292-2.5KG
BDH9292-500G
Sodium hydroxide ≥97.0%, beads ACS analytical reagent, VWR Chemicals BDH®
Sodium hydroxide
Meets reagent specifications for testing USP/NF monographs.
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
Calcium | <= 0.005 % |
Chloride | <= 0.005 % |
Heavy Metals | <= 0.002 % |
Iron | <= 0.001 % |
Magnesium | <= 0.002 % |
Mercury | <= 0.1 ppm |
Nickel | <= 0.001 % |
Nitrogen Compounds | <= 0.001 % |
Phosphate | <= 0.001 % |
Potassium | <= 0.02 % |
Purity | >= 97.0 % |
Sodium Carbonate | <= 1.0 % |
Sulfate | <= 0.003 % |
Meets reagent specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...