Sodium chloride ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Saline, NaCl, Sea salt, Halite
BDH9286-2.5KG
BDH9286-12KG
BDHVBDH9286-250LB
BDH9286-500G
BDH9286-50KG
BDH9286-2.5KGCS
591.68
CAD
BDH9286-2.5KG
BDH9286-12KG
BDH9286-250LB
BDH9286-500G
BDH9286-50KG
Sodium chloride ≥99.0% ACS, VWR Chemicals BDH®
Sodium chloride
Meets reagent specifications for testing USP/NF monographs.
White solid
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C (1013 hPa) Melting Pt: 801 °C Density: 2.16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,08599 |
Specification Test Results
Barium Test | PASS |
Bromide | <= 0.01 % |
Calcium | <= 0.002 % |
Chlorate & Nitrate | <= 0.003 % |
Heavy Metals | <= 0.0005 % |
Insolubles | <= 0.005 % |
Iodide | <= 0.002 % |
Iron | <= 0.0002 % |
Magnesium | <= 0.001 % |
pH (5%, Water) @25C | 5.0 - 9.0 |
Phosphate | <= 0.0005 % |
Potassium | <= 0.005 % |
Purity | >= 99.0 % |
Sulfate | <= 0.004 % |
Meets reagent specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...