Sodium acetate trihydrate 99.0-101.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Acetic acid sodium salt trihydrate
BDH9278-2.5KG
BDH9278-500G
BDH9278-2.5KGCS
1055.61
CAD
BDH9278-2.5KG
BDH9278-500G
Sodium acetate trihydrate 99.0-101.0% ACS analytical reagent, VWR Chemicals BDH®
Sodium acetate trihydrate
Meets reagent specifications for testing USP/NF monographs.
Colorless or white solid
Formula:
H₃CCOONa·3H₂O MW: 136.08 g/mol Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 58 Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Specification Test Results
Calcium |
|
Chloride |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Magnesium |
|
Permanganate Reducers Test | PASS |
pH (5%, Water) @25C | 7.5 - 9.2 |
Phosphate |
|
Potassium |
|
Purity | 99.0 - 101.0 % |
Sulfate |
|
Meets reagent specifications for testing USP/NF monographs. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...