Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

BDH9268-2.5KG BDH9268-500G
BDH9268-2.5KGCS 1290.17 CAD
BDH9268-2.5KG BDH9268-500G
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
Potassium dihydrogen phosphate

Meets reagent specifications for testing USP/NF monographs.


White or colorless solid

Formula: KH₂PO₄
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Chloride = 0.001 %
Heavy Metals = 0.001 %
Insolubles = 0.01 %
Iron = 0.002 %
Loss on Drying = 0.2 %
Nitrogen Compounds = 0.001 %
pH (5%, Water) @25C 4.1 - 4.5
Purity = 99.0 %
Sodium = 0.005 %
Sulfate = 0.003 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR