Potassium hydroxide ACS zA, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Caustic potash

BDH9262-500G BDH9262-2.5KG BDH9262-12KG
BDH9262-500GCS 937.02 CAD
BDH9262-500G BDH9262-2.5KG BDH9262-12KG
Potassium hydroxide ACS zA, VWR Chemicals BDH®
Potassium hydroxide

Meets reagent specifications for testing USP/NF monographs.


White, deliquescent solid

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Calcium = 0.005 %
Chloride = 0.01 %
Heavy Metals (as Ag) = 0.001 %
Iron = 0.001 %
Magnesium = 0.002 %
Nickel = 0.001 %
Nitrogen Compounds = 0.001 %
Phosphate = 5 ppm
Potassium Carbonate = 2.0 %
Purity = 85 %
Sodium = 0.05 %
Sulfate = 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR