Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Potassiumchloride

BDH9258-2.5KG BDH9258-500G
BDH9258-2.5KGCS 1074.29 CAD
BDH9258-2.5KG BDH9258-500G
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Potassium chloride

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Barium Test PASS
Bromide <= 0.01 %
Calcium <= 0.002 %
Chlorate & Nitrate (as NO3) <= 0.003 %
Heavy Metals <= 0.0005 %
Insolubles <= 0.005 %
Iodide <= 0.002 %
Iron <= 0.0003 %
Magnesium <= 0.001 %
pH (5%, Water) @25C 5.4 - 8.6
Phosphate <= 0.0005 %
Purity 99.0 - 100.5 %
Sodium <= 0.005 %
Sulfate <= 0.001 %
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR