Magnesium sulfate ≥98.0%, Reagent Grade, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: MgSO4Magnesium sulfate anhydrousMagnesium sulphate anhydrous

BDHVBDH9246-500G
BDH9246-500GCS 1694.52 CAD
BDH9246-500G
Magnesium sulfate ≥98.0%, Reagent Grade, VWR Chemicals BDH®
Magnesium sulphate

White powder.

Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
MDL Number: MFCD00011110
CAS Number: 7487-88-9
Merck Index: 12,05731
https://ca.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg

Order Now

Specification Test Results

Purity ≥98.0 %
Moisture ≤0.2 %
Appearance Fine white to off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR