L(+)-Ascorbic acid ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

BDH9242-100G BDH9242-250G
BDH9242-100GCS 2265.77 CAD
BDH9242-100G BDH9242-250G
L(+)-Ascorbic acid ACS analytical reagent, VWR Chemicals BDH®
L(+)-Ascorbic acid

White solid

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
Melting Pt: 190…192 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
REACH: exempt
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals (as Pb) = 0.002 %
Iron = 0.001 %
Purity = 99.0 %
Residue on Ignition = 0.1 %
Specific Rotation +20.5 - +21.5 degrees


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR