EDTA (ethylenediamine tetraacetic acid) ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Ethylenediaminetetraacetic acid2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acidOmniPur® EDTA(Ethylenedinitrilo)tetraacetic acidIdranal® IIKestranal® AEdetic acidEthylenedinitrilotetra-acetic acid

BDHVBDH9232-500G
BDH9232-500GCS 3702.49 CAD
BDH9232-500G
EDTA (ethylenediamine tetraacetic acid) ACS, VWR Chemicals BDH®
EDTA (Ethylenediamine tetraacetic acid)

White solid.

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00003541
CAS Number: 60-00-4
Merck Index: 13,03546
https://ca.vwr.com/stibo/web/std.lang.all/72/41/2297241.jpg

Order Now

Specification Test Results

Calcium = 0.001 %
Heavy Metals (as Pb) = 0.001 %
Insoluble in Dilute NH4OH = 0.005 %
Iron = 0.005 %
Magnesium = 0.0005 %
Nitrilotriacetic Acid = 0.1 %
Purity 99.4 - 100.6 %
Residue after Ignition = 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR