D-(+)-Glucose, anhydrous ACS, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Corn sugar
, D-Glucose
, D-(+)-Dextrose
, Glucolin
, ᴅ-(+)-Glucose
, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
, Glucose
, Dextrose
BDHVBDH9230-2.5KG
BDHVBDH9230-500G
BDH9230-500GCS
1147.39
CAD
BDH9230-500G
BDH9230-2.5KG
D-(+)-Glucose, anhydrous ACS, VWR Chemicals BDH®
D(+)-Glucose
Meets reagent specifications for testing USP/NF monographs.
White solid
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 146 °C Density: 1.54 g/cm³ (20 °C) |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 Merck Index: 14,04459 |
Specification Test Results
Chloride |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Loss on Drying |
|
Residue on Ignition |
|
Specific Rotation | +52.5 - +53.0 degrees |
Starch | PASS |
Sulfate & Sulfite |
|
Titrable Acid |
|
Meets reagent specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...