Citric acid monohydrate ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

BDH9228-2.5KG BDH9228-500G
BDH9228-2.5KGCS 1252.38 CAD
BDH9228-2.5KG BDH9228-500G
Citric acid monohydrate ACS analytical reagent, VWR Chemicals BDH®
Citric acid monohydrate

Colorless or white solid

Formula: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Melting Pt: 135…152 °C
Density: 1.552 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Carbonizable Substances PASS
Chloride = 0.001 %
Insolubles = 0.005 %
Iron = 3 ppm
Lead = 2 ppm
Oxalate Test PASS
Phosphate = 0.001 %
Purity 99.0 - 102.0 %
Residue after Ignition = 0.02 %
Sulfur Compounds (as SO4) = 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR