Orthoboric acid ≥99.5% ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

BDH9222-2.5KG
BDH9222-2.5KGCS 861.63 CAD
BDH9222-2.5KG
Orthoboric acid ≥99.5% ACS analytical reagent, VWR Chemicals BDH®
ortho-Boric acid

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium = 0.005 %
Chloride = 0.001 %
Heavy Metals (as Pb) = 0.001 %
Insolubles (Methanol) = 0.005 %
Iron = 0.001 %
Nonvolatiles (Methanol) = 0.05 %
Phosphate = 0.001 %
Purity = 99.5 %
Sulfate = 0.01 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR