Barium chloride dihydrate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Barium dichloride
BDHVBDH9220-500G
BDH9220-500GCS
1871.22
CAD
BDH9220-500G
Barium chloride dihydrate ≥99.0% ACS, VWR Chemicals BDH®
Barium chloride dihydrate
Meets reagent specifications for testing USP/NF monographs.
White solid
Formula:
BaCl₂·2H₂O MW: 244.27 g/mol Boiling Pt: 1560 °C (1013 hPa) Melting Pt: 960 °C Density: 3.86 g/cm³ (25 °C) |
MDL Number:
MFCD00149154 CAS Number: 10326-27-9 UN: 1564 ADR: 6.1,III Merck Index: 14,00971 |
Specification Test Results
Assay |
|
Calcium |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Loss on Drying | 14.0 - 16.0 % |
Oxidizing Substances (as NO3) |
|
pH (5%, Water) @25C | 5.2 - 8.2 |
Potassium |
|
Sodium |
|
Strontium |
|
Meets reagent specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...