Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®

Supplier: VWR International

BDH9216-2.5KG BDH9216-500G
BDH9216-2.5KGCS 976.14 CAD
BDH9216-2.5KG BDH9216-500G
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
Ammonium sulphate

Meets reagent specifications for testing USP/NF monographs.


Colorless powder or solid

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Chloride = 0.0005 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
Nitrate = 0.001 %
pH (5%, Water) @25C 5.0 - 6.0
Phosphate = 0.0005 %
Purity = 99.0 %
Residue on Ignition = 0.005 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR