Ammonium persulfate (APS) ≥98.0% ACS

Supplier: VWR International

Synonyms: Ammonium persulfatePeroxidisulphuric acid diammonium saltAmmonium persulphatePeroxidisulfuric acid diammonium salt

BDHVBDH9214-500G
BDH9214-500GCS 1455.33 CAD
BDH9214-500G
Ammonium persulfate (APS) ≥98.0% ACS
Ammonium peroxodisulphate (APS)

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid

Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
MDL Number: MFCD00003390
CAS Number: 7727-54-0
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://ca.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Order Now

Specification Test Results

Chloride & Chlorate = 0.001 %
Heavy Metals (as Pb) = 0.005 %
Insolubles = 0.005 %
Iron = 0.001 %
Manganese = 0.00005 %
Purity = 98.0 %
Residue on Ignition = 0.05 %
Titrable Free Acid = 0.04 meq/g
Meets reagent specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR