Ammonium chloride ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
BDH9208-2KG
BDH9208-500G
BDH9208-2KGCS
886.4
CAD
BDH9208-2KG
BDH9208-500G
Ammonium chloride ≥99.5% ACS, VWR Chemicals BDH®
Ammonium chloride
Meets reagent specifications for testing USP/NF monographs.
White solid
Formula:
NH₄Cl MW: 53.49 g/mol Melting Pt: 335 °C Density: 1.5256 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
Specification Test Results
Calcium | <= 0.001 % |
Heavy Metals (as Pb) | <= 0.0005 % |
Insolubles | <= 0.005 % |
Iron | <= 0.0002 % |
Magnesium | <= 0.0005 % |
pH (5%, Water) @25C | 4.5 - 5.5 |
Phosphate | <= 0.0002 % |
Purity | >= 99.5 % |
Residue on Ignition | <= 0.01 % |
Sulfate | <= 0.002 % |
Meets reagent specifications for testing USP/NF monographs. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...