Ammonium acetate ≥97.0% ACS analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Acetic acid ammonium salt

BDH9204-500G BDH9204-12KG BDH9204-2.5KG
BDH9204-500GCS 1368.45 CAD
BDH9204-500G BDH9204-12KG BDH9204-2.5KG
Ammonium acetate ≥97.0% ACS analytical reagent, VWR Chemicals BDH®
Ammonium acetate

Meets reagent specifications for testing USP/NF monographs.


Hygroscopic solid

Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride = 0.0005 %
Heavy Metals (as Pb) = 0.0005 %
Insolubles = 0.005 %
Iron = 0.0005 %
Nitrate = 0.001 %
pH (5%, Water) @25C 6.7 - 7.3
Purity = 97.0 %
Residue after Ignition = 0.01 %
Sulfate = 0.001 %
Meets reagent specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR