Stirrer Magnets with Raised Ring, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13655-25 13655-29 13655-31 13655-41 13655-37 13655-39 13655-33 13655-44
89077-872EA 102.64 CAD
89077-872 89077-874 89077-876 89077-878 89077-880 89077-882 89077-884 89077-886
Stirrer Magnets with Raised Ring, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Octagonal-shaped, PTFE stirrer magnets feature a large 25mm (1") pivot ring (PTFE collar).