Stirrer Magnets, X-Shaped , PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13560-01 13560-02 13560-03
89084-200EA 31.22 CAD
89084-200 89084-208 89084-214
Stirrer Magnets, X-Shaped , PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
Stirrer magnets feature an X-shaped vane with magnetic bar cross mounted
Designed for use with Microscale vials such as 89075-240 and 89075-266 (among others). Constructed of PTFE.