Stainless Steel Carts, Contec®
Supplier: Contec
2770 2769 2771
89237-614CS 1494.54 CAD
89237-614 89237-618 89237-622 89237-626
Stainless Steel Carts, Contec®
Mops
Stainless steel carts for use with 5 and 6.5 gal rectangular buckets with either heavy-duty or standard casters with brakes.
  • Lightweight and easy to maneuver
  • Resistant to most common chemicals and disinfectants
  • Heavy-duty option has locking casters
The carts are available with two different caster configurations (standard and heavy-duty) to cover a wide range of applications and environments. Designed to fit Contec's 5 or 6.5 gallon autoclavable plastic buckets which are offered in three colors (red, blue, and gray). Nonsterile. The carts are fullyMore Product Information