Sterile IPA 70% Bulk, ITW Texwipe
Supplier: Texwipe
TX3290 TX3290
CATWTX3290CS 2762.16 CAD
TWTX3290 CATWTX3290
Sterile IPA 70% Bulk, ITW Texwipe
Cleaning Agents Disinfectants
This Sterile 70% Isopropanol (IPA) contains 70% by volume USP-grade Isopropanol and 30% USP-purified water
Wide down for pass through to controlled environments.
For facilities currently blending and using isopropanol solutions.
To ensure sterility, this solution is submicron filtered, filled into cleaned containers, double-bagged and gamma-irradiated. Each bottle is lot coded with an expiration date for easy recordkeeping.
Certifications: IPA quality meets requirements of USP Isopropyl Alcohol. Water quality meets requirements of USP Purified Water.