Trubore® Bearing, Ace Glass Incorporated
Supplier: Ace Glass
9524-04 9524-06 9524-08
89075-046EA 165.52 CAD
89075-046 89075-048 89075-056
Trubore® Bearing, Ace Glass Incorporated
Stirrers Stirring Tools
Trubore® bearing with a standard taper joint for use with precision ground stir shafts
  • Match the bearing I.D. with stir shaft O.D. to select the proper shaft
  • Fabricated from borosilicate glass
The lubricant well at the top will accept enough Stir-Lube to provide hours of operation at up to 1000 rpm.