Tweezers, Angulated Tips, Style 6, Excelta Corp®
Supplier: Excelta
6-SA 6-SA-PI 6-SA-SE 6-S-SE 6-SA-PI-ET 6-SA-ET 6-SA-SE-ET
63042-756EA 78.49 CAD
63042-756 63042-760 63042-764 63042-766 63042-768 89411-792 89411-878 76471-482
Tweezers, Angulated Tips, Style 6, Excelta Corp®
Forceps
These unique angulated and sharp pointed tweezers are designed for manipulating very minute components or parts.
  • Uniquely angled for great accessability
  • Excellent visibility
  • Sharp pointed, exact and precise
The 'Style 6' Series tweezers feature 70° angled flat sharp precision pointed tips and can be utilized in a variety of applications, such as manipulating very minute components or parts. The angled tips allow for tweezing in restricted areas and enhance the visibility.We offer 3 different qualities. The higher the quality, the more precise the tips.More Product Information