Stopcock Plugs, Double Oblique Bore, Ace Glass Incorporated
Supplier: Ace Glass
8226-05 8226-08 8226-07 8226-09 8226-10
89062-784EA 132.53 CAD
89062-784 89062-786 89062-788 89062-780 89062-782
Stopcock Plugs, Double Oblique Bore, Ace Glass Incorporated
Stopcocks
These double oblique bore stopcocks are available with borosilicate glass or 1:5 PTFE plugs
The PTFE plugs (89062-784 and 89062-788) are designed for use with retainer O-Ring size –011.