Potassium methyl malonate 98%

Supplier: Thermo Scientific Chemicals
Warning

L20429-14 L20429-22
CAAAAL20429-14EA 102.38 CAD
CAAAAL20429-14 CAAAAL20429-22 CAAAL20429-36
Potassium methyl malonate 98%
Potassium methyl malonate

Purity: 98% CAS No.: 38330-80-2 Molecular Formula: CH3O2CCH2CO2K Formula Weight: 156.18 Melting Point: ca 190° dec. MDL No.: MFCD00014021

Formula: C₄H₅KO₄
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR