Samarium (III) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Samarium(III) trifluoromethanesulfonate, Samarium(III) trifluoromethanesulphonate

L20253-06 L20253-14
CAAAAL20253-06EA 99.54 CAD
CAAAAL20253-06 CAAAAL20253-14
Samarium (III) trifluoromethanesulfonate 98%
Samarium (III) trifluoromethanesulphonate

Purity: 98% CAS No.: 52093-28-4 Molecular Formula: Sm(SO3CF3)3 Formula Weight: 597.56 MDL No.: MFCD00192431

Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192431
CAS Number: 52093-28-4
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR