Dysprosium(III) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger

L20252-06 L20252-14
CAAAAL20252-06EA 118.82 CAD
CAAAAL20252-06 CAAAAL20252-14
Dysprosium(III) trifluoromethanesulfonate 98%
Dysprosium (III) trifluoromethanesulphonate
Formula: C₃DyF₉O₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00209583
CAS Number: 139177-62-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR