Cerium(III) trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Warning

L20251-06 L20251-14
CAAAAL20251-06EA 117.39 CAD
CAAAAL20251-06 CAAAAL20251-14
Cerium(III) trifluoromethanesulfonate 98%
Cerium (III) trifluoromethanesulphonate

Purity: 98% CAS No.: 76089-77-5 Molecular Formula: C3CeF9O9S3 Formula Weight: 587.33 Melting Point: 300° MDL No.: MFCD02093899

Formula: C₃CeF₉O₉S₃
MW: 587.33
Melting Pt: >300 °C
Density: 1.7
Storage Temperature: Ambient
MDL Number: MFCD02093899
CAS Number: 76089-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR