3-Buten-1-amine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20157-03
L20157-06
CAAAAL20157-03EA
229.1
CAD
CAAAAL20157-03
CAAAAL20157-06
3-Buten-1-amine 97%
3-Buten-1-amine
Purity: 97%
CAS No.: 2524-49-4
Molecular Formula: C4H9N
Formula Weight: 71.12
Flash Point: -5°(23°F)
Refractive Index: 1.4240
Odor: Amine-like
Boiling Point: 74-77°
Beilstein Reference: 1735803
UN No.: 2733
MDL No.: MFCD03425859
Formula:
C₄H₉N Boiling Pt: 74…77 °C Density: 0.777 Flash Pt: -5 °C(23 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03425859 CAS Number: 2524-49-4 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...