(R)-(+)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

L20079-03 L20079-06
CAAAAL20079-03EA 127.03 CAD
CAAAAL20079-03 CAAAAL20079-06 CAAAAL20079-14
(R)-(+)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®
(R)-(+)-4-Bromo-α-methylbenzylamine

Purity: 99% CAS No.: 45791-36-4 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Refractive Index: 1.5670 Melting Point: -25° (neat) Boiling Point: 140-145°/30mm Beilstein Reference: 2412319 UN No.: 2735 MDL No.: MFCD00066025

Formula: C₈H₁₀BrN
Boiling Pt: 140…145 °C/30 mm
Melting Pt: –25 °C
Density: 1.371
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00025548
CAS Number: 45791-36-4
EINECS: 256-245-0
UN: 2735
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR