Cerium(IV) trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20056-06
L20056-14
CAAAAL20056-06EA
84.69
CAD
CAAAAL20056-06
CAAAAL20056-14
Cerium(IV) trifluoromethanesulfonate 98%
Cerium (IV) trifluoromethanesulphonate
Purity: 98%
CAS No.: 107792-63-2
Molecular Formula: (CF3SO3)4Ce
Formula Weight: 736.39
MDL No.: MFCD03844746
Formula:
C₄CeF₁₂O₁₂S₄ Storage Temperature: Ambient |
MDL Number:
MFCD03844746 CAS Number: 107792-63-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...